4-Bromo-2-chlorophenylacetic acid, tech

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Bromo-2-chlorophenyl)acetic acid

OR313074-1G
APOSOR313074-1GEA 50.9 EUR
APOSOR313074-1G
4-Bromo-2-chlorophenylacetic acid, tech
4-Bromo-2-chlorophenylacetic acid
Formel: C₈H₆BrClO₂
Molecular Weight: 249,489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847409
CAS-Nummer: 916516-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR