Isothiazole-5-carboxylic acid
Lieferant: Apollo Scientific
OR313078-1G
APOSOR313078-1GEA
82.8
EUR
APOSOR313078-1G
Isothiazole-5-carboxylic acid
Isothiazole-5-carboxylic acid
Formel:
C₄H₃NO₂S Molecular Weight: 129.137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369964 CAS-Nummer: 10271-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...