4-Chlor-3-methoxybenzaldehyd

Lieferant: Apollo Scientific

OR313086-25G
APOSOR313086-25GEA 125 EUR
APOSOR313086-25G
4-Chlor-3-methoxybenzaldehyd
4-Chloro-3-methoxybenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07787490
CAS-Nummer: 13726-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR