3-Amino-1-n-boc-azepane

Lieferant: Apollo Scientific

OR313087-1G OR313087-5G
APOSOR313087-1GEA 0 EUR
APOSOR313087-1G APOSOR313087-5G
3-Amino-1-n-boc-azepane
3-Amino-1-n-boc-azepane
Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06738754
CAS-Nummer: 609789-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR