Methyl 2-(4-formylphenyl)acetate 98%

Lieferant: Apollo Scientific
Achtung

OR313093-5G
APOSOR313093-5GEA 120 EUR
APOSOR313093-5G
Methyl 2-(4-formylphenyl)acetate 98%
Methyl 2-(4-formylphenyl)acetate
Formel: C₁₀H₁₀O₃
Molecular Weight: 178.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17018734
CAS-Nummer: 96524-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR