Methyl-4-(bromomethyl)-3-chlorobenzoate

Lieferant: Apollo Scientific

OR313095-5G
APOSOR313095-5GEA 213 EUR
APOSOR313095-5G
Methyl-4-(bromomethyl)-3-chlorobenzoate
Methyl-4-(bromomethyl)-3-chlorobenzoate
Formel: C₉H₈BrClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10566565
CAS-Nummer: 74733-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR