7-Nitro-1H-indazol-5-yl acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Nitro-1H-indazol-5-yl ethanoate, 5-Acetoxy-7-nitro-1H-indazole
OR3135-100MG
APOSOR3135-100MGEA
325
EUR
APOSOR3135-100MG
7-Nitro-1H-indazol-5-yl acetate
7-Nitro-1H-indazol-5-yl acetate
Formel:
C₉H₇N₃O₄ Molecular Weight: 221,17 g/mol Schmelzpunkt: 193…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998771 CAS-Nummer: 1031927-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...