3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
Lieferant: Apollo Scientific
|
Achtung
|
OR3136-100MG
APOSOR3136-100MGEA
21.9
EUR
APOSOR3136-100MG
3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
Formel:
C₁₃H₁₆N₂O Molecular Weight: 216,29 g/mol Schmelzpunkt: 108…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051762 CAS-Nummer: 6631-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...