4'-Chloro-2-(2,4-dichlorophenyl)acetophenone
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(4-Chlorophenyl)-2-(2,4-dichlorophenyl)ethanone
OR314001-1G
APOSOR314001-1GEA
306
EUR
APOSOR314001-1G
4'-Chloro-2-(2,4-dichlorophenyl)acetophenone
4'-Chloro-2-(2,4-dichlorophenyl)acetophenone
Formel:
C₁₄H₉Cl₃O Molecular Weight: 299,58 g/mol Schmelzpunkt: 127…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16038495 CAS-Nummer: 654682-18-5 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...