2-(4-Chlorophenyl)-2',4'-dichloroacetophenone
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)ethanone
OR314002-1G
APOSOR314002-1GEA
383
EUR
APOSOR314002-1G
2-(4-Chlorophenyl)-2',4'-dichloroacetophenone
2-(4-Chlorophenyl)-2',4'-dichloroacetophenone
Formel:
C₁₄H₉Cl₃O Molecular Weight: 299,58 g/mol Schmelzpunkt: 65…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18452152 CAS-Nummer: 94171-11-6 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...