1-(3,5-Dichlorophenyl)ethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,5-Dichlorophenyl)ethylamine Hydrochloride

OR314005-1G OR314005-250MG OR314005-5G
APOSOR314005-1GEA 150 EUR
APOSOR314005-1G APOSOR314005-250MG APOSOR314005-5G
1-(3,5-Dichlorophenyl)ethylamine hydrochloride
1-(3,5-Dichlorophenyl)ethylamine hydrochloride
Formel: C₈H₉Cl₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18705305
CAS-Nummer: 321318-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR