Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-formyl-1-methyl-1H-pyrazole-5-carboxylate
OR314008-250MG
APOSOR314008-250MGEA
66.7
EUR
APOSOR314008-250MG
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate 97%
Methyl 3-formyl-1-methyl-1h-pyrazole-5-carboxylate
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22376784 CAS-Nummer: 1031351-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...