6-(Bromomethyl)pyridine-2-carbonitrile 95+%

Lieferant: Apollo Scientific

Synonyme: 6-(Bromomethyl)picolinonitrile, 6-Bromomethyl-2-cyanopyridine

OR314010-250MG
APOSOR314010-250MGEA 243 EUR
APOSOR314010-250MG
6-(Bromomethyl)pyridine-2-carbonitrile 95+%
6-(Bromomethyl)pyridine-2-carbonitrile
Formel: C₇H₅BrN₂
Molecular Weight: 197,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10687145
CAS-Nummer: 104508-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR