6-Bromo-2-methoxynicotinic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR314013-250MG
APOSOR314013-250MGEA
150
EUR
APOSOR314013-250MG
6-Bromo-2-methoxynicotinic acid 95%
6-Bromo-2-methoxynicotinic acid
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188730 CAS-Nummer: 1060806-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...