2-Bromo-6-chloro-pyridine-3-carbaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-6-chloronicotinaldehyde

OR314014-250MG
APOSOR314014-250MGEA 78.3 EUR
APOSOR314014-250MG
2-Bromo-6-chloro-pyridine-3-carbaldehyde 95+%
2-Bromo-6-chloro-pyridine-3-carbaldehyde
Formel: C₆H₃BrClNO
Molecular Weight: 220,451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11844804
CAS-Nummer: 1060815-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR