3-(Hydroxymethyl)-1h-pyrazole-5-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 3-(Hydroxymethyl)-1H-pyrazole-5-carboxylic acid

OR314039-250MG
APOSOR314039-250MGEA 231 EUR
APOSOR314039-250MG
3-(Hydroxymethyl)-1h-pyrazole-5-carboxylic acid 95+%
3-(Hydroxymethyl)-1h-pyrazole-5-carboxylic acid
Formel: C₅H₆N₂O₃
Molecular Weight: 142,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619746
CAS-Nummer: 1401555-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR