2,3,5-Trichloro-pyridin-4-ol 95+%

Lieferant: Apollo Scientific

Synonyme: 2,3,5-Trichloropyridin-4-ol

OR314052-250MG
APOSOR314052-250MGEA 224 EUR
APOSOR314052-250MG
2,3,5-Trichloro-pyridin-4-ol 95+%
2,3,5-Trichloro-pyridin-4-ol
Formel: C₅H₂Cl₃NO
Molecular Weight: 198,434 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01690819
CAS-Nummer: 1970-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR