(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (6-chloro-4-iodopyridin-3-yl)carbamate

OR314060-1G
APOSOR314060-1GEA 43.5 EUR
APOSOR314060-1G
(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester 98%
(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester
Formel: C₁₀H₁₂ClIN₂O₂
Molecular Weight: 354,572 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977418
CAS-Nummer: 400777-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR