6-Chloro-4-iodo-3-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

OR314078-1G
APOSOR314078-1GEA 77 EUR
APOSOR314078-1G
6-Chloro-4-iodo-3-hydroxypyridine
6-Chloro-4-iodo-3-hydroxypyridine
Formel: C₅H₃ClINO
Lagertemperatur: Raumtemperatur
MDL: MFCD11977445
CAS-Nummer: 877133-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR