Caronic anhydride

Lieferant: Apollo Scientific

Synonyme: 3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

OR315033-100G
APOSOR315033-100GEA 59.6 EUR
APOSOR315033-100G
Caronic anhydride
Caronic anhydride
Formel: C₇H₈O₃
Molecular Weight: 140.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09751198
CAS-Nummer: 67911-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR