Isochinolin-1-amin
Lieferant: Apollo Scientific
Synonyme:
Isoquinolin-1-amine, 1-Aminoisoquinoline
OR315041-5G
APOSOR315041-5GEA
68.3
EUR
APOSOR315041-5G
Isochinolin-1-amin
Isochinolin-1-amin
Formel:
C₉H₈N₂ Molecular Weight: 144.18 g/mol Siedepunkt: 324 Dichte: 1.21 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024137 CAS-Nummer: 1532-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...