3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-2H-pyrido[3,2-b]-1,4-oxazine

OR315044-1G
APOSOR315044-1GEA 0 EUR
APOSOR315044-1G
3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine
3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025907
CAS-Nummer: 20348-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR