10-Hydroxybenzo[h]quinoline 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR315046-25G
APOSOR315046-25GEA
144
EUR
APOSOR315046-25G
10-Hydroxybenzo[h]quinoline 98%
10-Hydroxybenzo[h]quinoline
Formel:
C₁₃H₉NO Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142350 CAS-Nummer: 33155-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...