4-Methyl-1-naphthoesäure
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-1-naphthalenecarboxylic acid
OR315051-100G
APOSOR315051-100GEA
85.7
EUR
APOSOR315051-100G
4-Methyl-1-naphthoesäure
4-Methyl-1-naphthoesäure
Formel:
C₁₂H₁₀O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671557 CAS-Nummer: 4488-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...