2-Chloro-6-methoxypyridin-3-amine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-chloro-6-methoxypyridine

OR315072-5G
APOSOR315072-5GEA 81.2 EUR
APOSOR315072-5G
2-Chloro-6-methoxypyridin-3-amine 98%
2-Chloro-6-methoxypyridin-3-amine
Formel: C₆H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09258809
CAS-Nummer: 34392-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR