3-Iodo-2-nitropyridine 98%
Lieferant: Apollo Scientific
OR315076-5G
APOSOR315076-5GEA
92.8
EUR
APOSOR315076-5G
3-Iodo-2-nitropyridine 98%
3-Iodo-2-nitropyridine
Formel:
C₅H₃IN₂O₂ Molecular Weight: 249.994 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08061318 CAS-Nummer: 54231-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...