3-Chloro-2-methylpyridine

Lieferant: Apollo Scientific

OR315077-1G
APOSOR315077-1GEA 157 EUR
APOSOR315077-1G
3-Chloro-2-methylpyridine
3-Chloro-2-methylpyridine
Formel: C6H6ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD04114121
CAS-Nummer: 72093-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR