3-Bromo-5-ethylpyridine 98%

Lieferant: Apollo Scientific

OR315080-1G
APOSOR315080-1GEA 120 EUR
APOSOR315080-1G
3-Bromo-5-ethylpyridine 98%
3-Bromo-5-ethylpyridine
Formel: C7H8BrN
Molecular Weight: 186,0491 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800592
CAS-Nummer: 142337-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR