5-Amino-2,3-dichlorpyridin 98%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dichlorpyridin-5-amin, 2,3-Dichlor-5-pyridylamin
OR315086-25G
APOSOR315086-25GEA
150
EUR
APOSOR315086-25G
5-Amino-2,3-dichlorpyridin 98%
5-Amino-2,3-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840434 CAS-Nummer: 98121-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...