Methyl 4-methylnicotinate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-methylnicotinate
OR315098-1G
OR315098-10G
OR315098-5G
APOSOR315098-1GEA
78.3
EUR
APOSOR315098-1G
APOSOR315098-10G
APOSOR315098-5G
Methyl 4-methylnicotinate 98%
Methyl-4-methylnicotinate
Formel:
C₈H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258869 CAS-Nummer: 33402-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...