Ethyl(pyridin-2-ylmethyl)amine 97%

Lieferant: Apollo Scientific

Synonyme: N-(Pyridin-2-ylmethyl)ethanamine

OR315116-5G
APOSOR315116-5GEA 52.2 EUR
APOSOR315116-5G
Ethyl(pyridin-2-ylmethyl)amine 97%
Ethyl(pyridin-2-ylmethyl)amine
Formel: C₈H₁₂N₂
Molecular Weight: 136,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07411568
CAS-Nummer: 51639-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR