2,3-Dibromo-5,6-dimethylpyridine

Lieferant: Apollo Scientific

OR315119-1G
APOSOR315119-1GEA 60.9 EUR
APOSOR315119-1G
2,3-Dibromo-5,6-dimethylpyridine
2,3-Dibromo-5,6-dimethylpyridine
Formel: C₇H₇Br₂N
Molecular Weight: 264,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13688994
CAS-Nummer: 117846-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR