3-Amino-2-methoxy-4-picolin 95+%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxy-4-methylpyridin-3-amine
OR315127-1G
APOSOR315127-1GEA
88.6
EUR
APOSOR315127-1G
3-Amino-2-methoxy-4-picolin 95+%
3-Amino-2-methoxy-4-picolin
Formel:
C₇H₁₀N₂O Molecular Weight: 138,1671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277287 CAS-Nummer: 76005-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...