5-Brom-3-chlor-2-methoxypyridin 98%

Lieferant: Apollo Scientific

OR315131-5G
APOSOR315131-5GEA 55.1 EUR
APOSOR315131-5G
5-Brom-3-chlor-2-methoxypyridin 98%
5-Brom-3-chlor-2-methoxypyridin
Formel: C₆H₅BrClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04972382
CAS-Nummer: 848366-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR