(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%

Lieferant: Apollo Scientific

OR315137-250MG
APOSOR315137-250MGEA 178 EUR
APOSOR315137-250MG
(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%
(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165.618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09749902
CAS-Nummer: 359849-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR