4-Picolylbromid Hydrobromid 95%
Lieferant: Apollo Scientific
Synonyme:
4-(Bromomethyl)pyridine hydrobromide
OR315149-25G
APOSOR315149-25GEA
78.3
EUR
APOSOR315149-25G
4-Picolylbromid Hydrobromid 95%
4-Picolylbromid Hydrobromid
Formel:
C₆H₇Br₂N Molecular Weight: 252,94 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863545 CAS-Nummer: 73870-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...