2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(E)-Methyl 2-(2-((6-chloropyrimidin-4-yl)oxy)phenyl)-3-methoxyacrylate
OR315151-100G
APOSOR315151-100GEA
163
EUR
APOSOR315151-100G
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester
Formel:
C₁₅H₁₃ClN₂O₄ Molecular Weight: 320,728 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13194671 CAS-Nummer: 131860-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...