9-Phenylcarbazole-2-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (9-Phenyl-9H-carbazol-2-yl)boronic acid

OR315156-25G
APOSOR315156-25GEA 112 EUR
APOSOR315156-25G
9-Phenylcarbazole-2-boronic acid
9-Phenylcarbazole-2-boronic acid
Formel: C₁₈H₁₄BNO₂
Molecular Weight: 287.13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22207050
CAS-Nummer: 1001911-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR