4-(9-Carbazolyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 4-(9H-9-carbozale)phenylboronic acid, 4-(9H-Carbazol-9-yl)phenylboronic acid, (4-(9H-Carbazol-9-yl)phenyl)boronic acid, 4-(9H-Carbazol-9-yl)benzeneboronic acid

OR315164-25G
APOSOR315164-25GEA 89.9 EUR
APOSOR315164-25G
4-(9-Carbazolyl)benzeneboronic acid 98%
4-(9-Carbazolyl)benzeneboronic acid
Formel: C₁₈H₁₄BNO₂
Molecular Weight: 287,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176534
CAS-Nummer: 419536-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR