H-Aib-OtBu·HCl 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 2-aminoisobutyrate hydrochloride, tert-Butyl 2-amino-2-methylpropanoate hydrochloride
OR315173-100G
APOSOR315173-100GEA
200
EUR
APOSOR315173-100G
H-Aib-OtBu·HCl 97%
H-Aib-OtBu·HCl
Formel:
C₈H₁₇NO₂·HCl Molecular Weight: 195,7 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08272284 CAS-Nummer: 84758-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...