(R)-(+)-3-Amino-1,2-propandiol 95+%
Lieferant: Apollo Scientific
OR315176-25G
APOSOR315176-25GEA
97.3
EUR
APOSOR315176-25G
(R)-(+)-3-Amino-1,2-propandiol 95+%
(R)-(+)-3-Amino-1,2-propandiol
Formel:
C₃H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798261 CAS-Nummer: 66211-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...