Spiro[2.5]oct-1-ylamine hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315179-250MG
APOSOR315179-250MGEA
449
EUR
APOSOR315179-250MG
Spiro[2.5]oct-1-ylamine hydrochloride 95%
Spiro[2.5]oct-1-ylamine hydrochloride
Formel:
C₈H₁₅N·ClH Molecular Weight: 161,672 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23144279 CAS-Nummer: 17202-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...