4-Bromo-5-thiazolecarboxylic acid ethyl ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-bromothiazole-5-carboxylate, Ethyl 4-bromo-5-thiazolecarboxylate

OR315188-1G
APOSOR315188-1GEA 139 EUR
APOSOR315188-1G
4-Bromo-5-thiazolecarboxylic acid ethyl ester 98%
4-Bromo-5-thiazolecarboxylic acid ethyl ester
Formel: C₆H₆BrNO₂S
Molecular Weight: 236.09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09909677
CAS-Nummer: 152300-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR