N-(5-Bromothiazol-2-yl)acetamide hydrobromide 95+%

Lieferant: Apollo Scientific

OR315192-1G OR315192-25G OR315192-5G
APOSOR315192-1GEA 191 EUR
APOSOR315192-1G APOSOR315192-25G APOSOR315192-5G
N-(5-Bromothiazol-2-yl)acetamide hydrobromide 95+%
N-(5-Bromothiazol-2-yl)acetamide hydrobromide
Formel: C₅H₅BrN₂OS·BrH
Molecular Weight: 301.987 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20921811
CAS-Nummer: 1354359-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR