2,5-Pyrazinedicarboxylic acid dihydrate 95%

Lieferant: Apollo Scientific
Achtung

OR315199-5G
APOSOR315199-5GEA 132 EUR
APOSOR315199-5G
2,5-Pyrazinedicarboxylic acid dihydrate 95%
2,5-Pyrazinedicarboxylic acid dihydrate
Formel: C₆H₆N₂O₅
Molecular Weight: 186,123 g/mol
Schmelzpunkt: 272…277 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216919
CAS-Nummer: 205692-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR