2-Brom-5-iod-4-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Brom-5-iod-4-picolin

OR315201-1G
APOSOR315201-1GEA 92.8 EUR
APOSOR315201-1G
2-Brom-5-iod-4-methylpyridin 97%
2-Bromo-5-iodo-4-methylpyridine
Formel: C₆H₅BrIN
Molecular Weight: 297,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037465
CAS-Nummer: 942206-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR