2,4-Dibrom-5-nitropyridin 98%

Lieferant: Apollo Scientific
Achtung

OR315206-1G
APOSOR315206-1GEA 94.4 EUR
APOSOR315206-1G
2,4-Dibrom-5-nitropyridin 98%
2,4-Dibromo-5-nitropyridine
Formel: C₅H₂Br₂N₂O₂
Molecular Weight: 281,8900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832226
CAS-Nummer: 4487-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR