4-Chlorpyridin-N-oxid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315222-1G
OR315222-5G
OR315222-25G
APOSOR315222-1GEA
79.9
EUR
APOSOR315222-1G
APOSOR315222-5G
APOSOR315222-25G
4-Chlorpyridin-N-oxid 95%
4-Chlorpyridin-N-oxid
Formel:
C₅H₄ClNO Schmelzpunkt: 160 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00047425 CAS-Nummer: 1121-76-2 EINECS: 214-336-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...