5-Norbornene-2-carboxaldehyde
Lieferant: Apollo Scientific
Synonyme:
Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde
OR315246-100G
APOSOR315246-100GEA
154
EUR
APOSOR315246-100G
5-Norbornene-2-carboxaldehyde
5-Norbornene-2-carboxaldehyde
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167568 CAS-Nummer: 5453-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...