2,6-Diethylphenylisothiocyanat

Lieferant: Apollo Scientific

Synonyme: 1,3-Diethyl-2-isothiocyanatobenzene

OR315247-5G
APOSOR315247-5GEA 132 EUR
APOSOR315247-5G
2,6-Diethylphenylisothiocyanat
2,6-Diethylphenylisothiocyanat
Formel: C₁₁H₁₃NS
Molecular Weight: 191,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041069
CAS-Nummer: 25343-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR